Sartorius™ Air Filter
Overview
Brand: Sartorius™
Manufacturer Part Number: 17804--------NPG
X25 MIDISART 0,45µM 1/8MNPTStck.)
UNSPSC: 41000000
Code: NEW
Additional Details:
Additional Details: Weight: 0.10000kg
Product Code. 10706672
Quantity Price
1 £ 153.0 / Pack of 25
Estimated Shipment
27-02-2017
Add to basket
Description and Specification
Specification
Filtration Area (Metric) 20cm2
Autoclavable Yes
Sterile Yes
Type Air Filter
Quantity 25/pk
Housing Material Polypropylene
Filter Type Flat Disc Capsules
Pore Size 0.45μm