missing translation for 'onlineSavingsMsg'
Learn More

AIDA, Tocris Bioscience™

Potent and selective group I mGlu antagonist

Brand:  Tocris (Bio-Techne) 0904/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13468133

  • £182.28 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A relatively potent and selective antagonist of group I metabotropic glutamate receptors (mGlu1a), having no effect on group II (mGlu2) or group III (mGlu4) receptors expressed individually in baby hamster kidney cells. Has no effect on ionotropic glutamate receptors. Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

168560-79-0
221.212
aida, 1-aminoindan-1,5-dicarboxylic acid, gnf-pf-1401, 1-amino-2,3-dihydro-1h-indene-1,5-dicarboxylic acid, 1-aminoindan-1, 5-dicarboxylic acid, 1h-indene-1,5-dicarboxylic acid, 1-amino-2,3-dihydro, indan-1,5-dicarboxylic acid, 1-amino, rs-1-aminoindan-1,5-dicarboxylic acid, aida, solid, aidc
1-amino-2,3-dihydroindene-1,5-dicarboxylic acid
C11H11NO4
LSTPKMWNRWCNLS-UHFFFAOYSA-N
2071
C1CC(C2=C1C=C(C=C2)C(=O)O)(C(=O)O)N

Specifications

10mg
168560-79-0
aida, 1-aminoindan-1,5-dicarboxylic acid, gnf-pf-1401, 1-amino-2,3-dihydro-1h-indene-1,5-dicarboxylic acid, 1-aminoindan-1, 5-dicarboxylic acid, 1h-indene-1,5-dicarboxylic acid, 1-amino-2,3-dihydro, indan-1,5-dicarboxylic acid, 1-amino, rs-1-aminoindan-1,5-dicarboxylic acid, aida, solid, aidc
C1CC(C2=C1C=C(C=C2)C(=O)O)(C(=O)O)N
221.212
221.21
AIDA
C11H11NO4
LSTPKMWNRWCNLS-UHFFFAOYSA-N
1-amino-2,3-dihydroindene-1,5-dicarboxylic acid
2071
>98%
Videos
Documents