Learn More
AIDA, Tocris Bioscience™
Potent, selective group I mGlu antagonist
Brand:  Tocris (Bio-Techne) 090450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488133
  • £608.00 / 50mg
Estimated Shipment
28-06-2018
Add to basket
Description
Description
A relatively potent and selective antagonist of group I metabotropic glutamate receptors (mGlu1a), having no effect on group II (mGlu2) or group III (mGlu4) receptors expressed individually in baby hamster kidney cells. Has no effect on ionotropic glutamate receptors. Centrally active following systemic administration in vivo.
Specifications
Chemical Identifiers
AIDA
C11H11NO4
221.21
(RS)-1-Aminoindan-1, 5-dicarboxylic acid
Specifications
AIDA
>98%
221.21
(RS)-1-Aminoindan-1, 5-dicarboxylic acid
168560-79-0
C11H11NO4