missing translation for 'onlineSavingsMsg'
Learn More

AHR Monoclonal Antibody (4MEJJ), PE-Cyanine7, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-5925-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15588596

  • £140.00 / 25µg
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The aryl hydrocarbon receptor (AhR), also known as the dioxin receptor, is a ligand-activated helix/loop/helix transcription factor found in a variety of vertebrate species. The known ligands for AhR are foreign planar aromatic compounds, such as polycyclic aromatic compounds and halogenated aromatic compounds such as 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD). Unlike the steroid/thyroid hormone receptors, there is no known physiological ligand for the Ah Receptor. Studies indicate that in non-ligand activated cells, AhR is found complexed with HSP90 predominantly in the cytoplasm. Upon binding to an agonist, the ligand-activated AhR is believed to transform to a nuclear, DNA binding form. This transformation process appears to involve dissociation of HSP90 from AhR followed by formation of a heterodimer with AhR nuclear translocator protein (Arnt). The AhR-ligand complex appears to initiate gene transcription of cytochrome P450 1A1.

AHR Monoclonal antibody specifically detects AHR in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

AHR
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P30561
AHR
E. coli derived, His-tagged AHR (aa 443-805)
25 μg
Primary
Mouse
Antibody
IgG2a κ
Flow Cytometry
4MEJJ
PE-Cyanine7
AHR
Ah; Ah receptor; Ahh; Ahr; Ahre; AH-receptor; aromatic hydrocarbon receptor; aryl hydrocarbon receptor; aryl-hydrocarbon receptor; bHLHe76; Class E basic helix-loop-helix protein 76; dioxin receptor; In
Rat
Affinity chromatography
RUO
11622
4° C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.