missing translation for 'onlineSavingsMsg'
Learn More

AHR Rat anti-Mouse, PE, Clone: 4MEJJ, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5925-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557056

  • £262.30 / 100µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

This 4MEJJ monoclonal antibody recognizes mouse aryl hydrocarbon receptor (AHR). The AHR is a ligand-activated transcription factor that mediates the toxic effects of a diverse group of environmental contaminants, most notably aryl hydrocarbons such as polychlorinated biphenyls (PCB) and tetrachlorodibenzo-p-dioxin (TCDD). The AHR has also been shown to bind to a number of naturally occurring compounds found in fruits and vegetables as well as compounds generated through normal cellular metabolism. AHR is localized in the cytoplasm in a complex that includes HSP90, p23, and XAP2/AIP/ARA9. Upon ligand-binding, AHR translocates to the nucleus and binds with aryl hydrocarbon receptor nuclear translocator (ARNT), and this complex binds to the consensus DNA sequence, GCGTG, found in the promoter/enhancer regions of many genes such as CYP1A1. The AHR is expressed in many cell types, with highest expression levels found in liver. The AHR has been shown to play a role in the regulation/differentiation of Treg and Th17 cells.

The 4MEJJ monoclonal antibody has been found to recognize the b-1, b-2 and d alleles of mouse AHR but does not crossreact with human AHR.
Specifications

Specifications

AHR
4MEJJ
PE
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Aryl hydrocarbon receptor
IgG2a
RUO
Primary
Murine