missing translation for 'onlineSavingsMsg'
Learn More

AHR Monoclonal Antibody (4MEJJ), eFluor™ 660, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

£138.00 - £323.00

Specifications

Antigen AHR
Clone 4MEJJ
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15599986
View Documents Promotion Details
Affymetrix eBioscience
50-5925-80
25 μg
£138.00
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15509996
View Documents Promotion Details
Affymetrix eBioscience
50-5925-82
100 μg
£323.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: This 4MEJJ monoclonal antibody recognizes mouse aryl hydrocarbon receptor (AHR). The AHR is a ligand-activated transcription factor that mediates the toxic effects of a diverse group of environmental contamits, most notably aryl hydrocarbons such as polychlorinated biphenyls (PCB) and tetrachlorodibenzo-p-dioxin (TCDD). The AHR has also been shown to bind to a number of naturally occurring compounds found in fruits and vegetables as well as compounds generated through normal cellular metabolism. AHR is localized in the cytoplasm in a complex that includes HSP90, p23, and XAP2/AIP/ARA9. Upon ligand-binding, AHR translocates to the nucleus and binds with aryl hydrocarbon receptor nuclear translocator (ARNT), and this complex binds to the consensus DNA sequence, GCGTG, found in the promoter/enhancer regions of many genes such as CYP1A1. The AHR is expressed in many cell types, with highest expression levels found in liver. The AHR has been shown to play a role in the regulation/differentiation of Treg and Th17 cells. The 4MEJJ monoclonal antibody has been found to recognize the b-1, b-2 and d alleles of mouse AHR but does not crossreact with human AHR. Applications Reported: This 4MEJJ antibody has been reported for use in intracellular staining followed by flow cytometric analysis.

AHR Monoclonal antibody specifically detects AHR in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

AHR
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
Ah; Ah receptor; Ahh; Ahr; Ahre; AH-receptor; aromatic hydrocarbon receptor; aryl hydrocarbon receptor; aryl-hydrocarbon receptor; bHLHe76; Class E basic helix-loop-helix protein 76; dioxin receptor; In
AHR
IgG2a κ
Affinity chromatography
Antibody
4MEJJ
Flow Cytometry
eFluor 660
Rat
Mouse
P30561
11622
E. coli derived, His-tagged AHR (aa 443-805)
Primary
4° C, store in dark, DO NOT FREEZE!
AHR
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.