missing translation for 'onlineSavingsMsg'
Learn More

AHR, PE-Cyanine7, clone: FF3399, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-9854-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15588676

  • £113.00 / 25 tests

Estimated Shipment
28-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The FF3399 monoclonal antibody recognizes human aryl hydrocarbon receptor (AHR). The AHR is a ligand-activated transcription factor that mediates the toxic effects of a diverse group of environmental contaminants, most notably aryl hydrocarbons such as polychlorinated biphenyls (PCB) and tetrachlorodibenzo-p-dioxin (TCDD). The AHR has also been shown to bind to a number of naturally occurring compounds found in fruits and vegetables as well as compounds generated through normal cellular metabolism. AHR is localized in the cytoplasm in a complex that includes HSP90, p23, and XAP2/AIP/ARA9. Upon ligand-binding, AHR translocates to the nucleus and binds with aryl hydrocarbon receptor nuclear translocator (ARNT), and this complex binds to the consensus DNA sequence, GCGTG, found in the promoter/enhancer regions of many genes such as CYP1A1. The AHR is expressed in many cell types, with highest expression levels found in liver. The AHR has been shown to play a role in the regulation/differentiation of Treg and Th17 cells.Applications Reported: This FF3399 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This FF3399 antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of Th17-polarized normal human peripheral blood cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This antibody can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

AHR
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P35869
Ah receptor, AhR, Class E basic helix-loop-helix protein 76, bHLHe76, AHR, BHLHE76
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
FF3399
PE-Cyanine7
AHR
Liquid
AHR
IgG2b, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
196
Videos
Documents

For Research Use Only.