Learn More
AHNAK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553912
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DIKSPSLDVT VPEAELNLET PEISVGGKGK KSKFKMPKIH MSGPKIKAKK QGFDLNVPGG EIDASLKAPD VDVNIAGPDA ALKVDVKSPK TKKTMFGKMY FPDVEFDIKS PKFKAEAPLP SPKL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.
AHNAK Polyclonal antibody specifically detects AHNAK in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
AHNAK | |
Polyclonal | |
Unconjugated | |
AHNAK | |
AHNAK; AHNAK 1; AHNAK nucleoprotein; AHNAK nucleoprotein (desmoyokin); AHNAK-related; AHNAKRS; Desmoyokin; DY6; MGC5395; Neuroblast differentiation-associated protein AHNAK; PM227; RGD1308572; UV118 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q09666 | |
AHNAK | |
Recombinant protein corresponding to Human AHNAK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only