Learn More
AHA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551480
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PTMNGESVDP VGQPALKTEE RKAKPAPSKT QARPVGVKIP TCKITLKETF LTSPEELYRV FTTQELVQAF THAPATLEAD RGGKFHMVDG NVSGEFTDLV PEKHIVMKWR FKSWPEGHFA TITLTFIDKN GETELCMEGR GIPAPEEE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
AHA1 Polyclonal antibody specifically detects AHA1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
AHA1 | |
Polyclonal | |
Unconjugated | |
Ahsa1 | |
Activator of 90 kDa heat shock protein ATPase homolog 1; activator of Hsp90 ATPase activity 1; Activator of Hsp90 ATPase protein 1; Aha 1; AHA1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1; AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast); AHA1, activator of heat shock protein ATPase 1; AHA1, activator of heat shock protein ATPase homolog 1; Aha1p; AHSA 1; Ahsa1; BC023857; C14orf3; hAha1; Hsp90 co-chaperone AHA1; HSPC322; p38; SHSA1; YD8142.16; YD8142B.06; YDR214W | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10598, 217737, 681996 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95433, Q8BK64 | |
Ahsa1 | |
Recombinant protein corresponding to Human AHA1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only