missing translation for 'onlineSavingsMsg'
Learn More

AH 6809, Tocris Bioscience™

EP1 and EP2 receptor antagonist

Brand:  RD Systems (Bio-Techne) 67150

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13437873

  • £571.28 / 50mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Antagonist at prostaglandin EP1 (pA2 = 6.8) and EP2 (Ki = 350 nM) receptors. Also weakly inhibits DP receptors (pA2 = 4.45).
Specifications

Chemical Identifiers

33458-93-4
298.294
6-isopropoxy-9-oxoxanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo-9h-xanthene-2-carboxylic acid, 6-isopropoxy-9-oxo-9h-xanthene-2-carboxylic acid, 9h-xanthene-2-carboxylicacid, 6-1-methylethoxy-9-oxo, 9h-xanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo, tocris-0671, d0j9pg, 6-isopropoxy-9-xanthone-2-carboxylic acid, ah, crystalline solid or supercooled liquid
9-oxo-6-propan-2-yloxyxanthene-2-carboxylic acid
C17H14O5
AQFFXPQJLZFABJ-UHFFFAOYSA-N
119461
CC(C)OC1=CC2=C(C=C1)C(=O)C3=C(O2)C=CC(=C3)C(=O)O

Specifications

AH 6809
33458-93-4
6-isopropoxy-9-oxoxanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo-9h-xanthene-2-carboxylic acid, 6-isopropoxy-9-oxo-9h-xanthene-2-carboxylic acid, 9h-xanthene-2-carboxylicacid, 6-1-methylethoxy-9-oxo, 9h-xanthene-2-carboxylic acid, 6-1-methylethoxy-9-oxo, tocris-0671, d0j9pg, 6-isopropoxy-9-xanthone-2-carboxylic acid, ah, crystalline solid or supercooled liquid
CC(C)OC1=CC2=C(C=C1)C(=O)C3=C(O2)C=CC(=C3)C(=O)O
298.294
298.3
50mg
C17H14O5
AQFFXPQJLZFABJ-UHFFFAOYSA-N
9-oxo-6-propan-2-yloxyxanthene-2-carboxylic acid
119461
>98%
Videos
Documents