missing translation for 'onlineSavingsMsg'
Learn More

AH 11110 hydrochloride, Tocris Bioscience™

Subtype-selective α1B ligand

Brand:  Tocris (Bio-Techne) 099810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13468253

  • £94.08 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A subtype-selective ligand for the α1B-adrenoceptor. The pKi values for binding to rat fibroblast cell membranes expressing α1B (hamster), α1A (bovine) and α1D (rat) are 7.1, 5.59, and 5.68 respectively. Appears to be non-selective between α1 subtypes and α2 receptors in smooth muscle preparations.
Specifications

Chemical Identifiers

179388-65-9
374.909
ah 11110 hydrochloride, 1-biphenyl-2-yloxy-4-imino-4-piperidin-1-yl-butan-2-ol hydrochloride, d0fh7b, ah hplc , solid, 4-imino-1-2-phenylphenoxy-4-piperidinebutan-2-ol hydrochloride, 4-imino-1-2-phenylphenoxy-4-piperidin-1-ylbutan-2-ol hydrochloride, 1-1,1'-biphenyl-2-yloxy-4-imino-4-piperidin-1-yl butan-2-ol hydrochloride
4-imino-1-(2-phenylphenoxy)-4-piperidin-1-ylbutan-2-ol;hydrochloride
C21H27ClN2O2
WWHGFIXWBLHHQB-UHFFFAOYSA-N
9907616
C1CCN(CC1)C(=N)CC(COC2=CC=CC=C2C3=CC=CC=C3)O.Cl

Specifications

10mg
179388-65-9
ah 11110 hydrochloride, 1-biphenyl-2-yloxy-4-imino-4-piperidin-1-yl-butan-2-ol hydrochloride, d0fh7b, ah hplc , solid, 4-imino-1-2-phenylphenoxy-4-piperidinebutan-2-ol hydrochloride, 4-imino-1-2-phenylphenoxy-4-piperidin-1-ylbutan-2-ol hydrochloride, 1-1,1'-biphenyl-2-yloxy-4-imino-4-piperidin-1-yl butan-2-ol hydrochloride
C1CCN(CC1)C(=N)CC(COC2=CC=CC=C2C3=CC=CC=C3)O.Cl
374.909
374.91
AH 11110 hydrochloride
C21H27ClN2O2
WWHGFIXWBLHHQB-UHFFFAOYSA-N
4-imino-1-(2-phenylphenoxy)-4-piperidin-1-ylbutan-2-ol;hydrochloride
9907616
>98%
Videos
Documents