missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human AGTR-2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | AGTR-2 |
---|---|
Clone | 364805 |
Concentration | LYOPH |
Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Description
AGTR-2 Monoclonal specifically detects AGTR-2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
AGTR-2 | |
LYOPH | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
P50052 | |
186 | |
HEK293 human embryonic kidney cell line transfected with human AGTR-2 Met1-Ser363 Accession # P50052 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human AGTR-2. Stains human AGTR-2-transfected cells but not irrelevant transfectants. |
364805 | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AGTR2, Angiostensin Receptor, angiotensin II receptor, type 2, Angiotensin II type-2 receptor, angiotensin receptor 2, AT2, AT2ATGR2, AT2R, MRX88, MRX88type-2 angiotensin II receptor | |
AGTR2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only