missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agrin Mouse anti-Rat, Clone: 76805, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Agrin |
---|---|
Clone | 76805 |
Immunogen | S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Agrin Ala1153-Pro1959 (Pro1788-Ser1798 del) Accession # P25304 |
Gene ID (Entrez) | 375790 |
Gene Accession No. | P25304 |
Description
Agrin Monoclonal specifically detects Agrin in Rat samples. It is validated for Western Blot.Specifications
Agrin | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat Agrin Ala1153-Pro1959 (Pro1788-Ser1798 del) Accession # P25304 | |
P25304 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Mouse | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
76805 | |
375790 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects rat Agrin in direct ELISAs and Western blots. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only