missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGBL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558201
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GNLGRVDQVS EFEYDLFIRP DTCNPRFRVW FNFTVENVKE SQRVIFNIVN FSKTKSLYRD GMAPMVKSTS RPKWQRLPPK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 38%.
AGBL4 Polyclonal antibody specifically detects AGBL4 in Human samples. It is validated for Immunohistochemistry.Specifications
AGBL4 | |
Polyclonal | |
Unconjugated | |
AGBL4 | |
AGBL4; ATP/GTP binding protein like 4; ATP/GTP binding protein-like 4; ATP/GTP-binding protein-like 4; CCP6; Cytosolic carboxypeptidase 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VU57 | |
AGBL4 | |
Recombinant protein corresponding to Human AGBL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only