Learn More
Retsch™ Agate Grinding Jars
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Agate Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand:  Retsch™ 01.462.0008
Code : A0
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 10771581
  • £665.33 / Each
Estimated Shipment
01-03-2018
Add to basket
Specifications
Specifications
10mL
Agate
For MM 200 Mixer Mill
Grinding Jar
Videos
View accessory products