Learn More
RETSCH Agate Grinding Jars
Use these grinding jars for dry, wet and cryogenic grinding of small samples. Retsch™ Agate Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand: RETSCH 01.462.0233
Code : 50
Additional Details : Weight : 0.30000kg
Description
- Screw-top grinding jars
- Available in a variety of sizes
- For use with Retsch mills
Specifications
Grinding Jar | |
10 mL | |
For MM 301 Mixer mill | |
Agate |