missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Agate Comfort Grinding Jars

Use these grinding jars with Retsch™ ball mills. Retsch™ Agate Comfort Grinding Jars feature an O-type sealing for safe operation.

Brand:  RETSCH 01.462.0136

Code : 50

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 10608321

  • £2170.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Available in a variety of sizes from 12 to 500mL
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications

Specifications

Comfort Grinding Jar
125 mL
For PM 100, PM 200, PM 400 and S 100
Agate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers