missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose, Standard, Low Electroendosmosis (EEO), Ultrapure Bioreagent, J.T. Baker™
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Description
- For Electrophoresis
Specifications
Quantity | 100 g |
Packaging | Poly Bottle |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | Agarose, Standard, Low Electroendosmosis (EEO) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'