missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Agarose Beads 6% B

Brand:  Alfa Aesar™ H32491.AU

UNSPSC : 41101902

Code : NEW

Additional Details : CAS Number : 9012-36-6 Weight : 0.00100kg Transport : UN number : 1170 Chem class : 3 Pack group : III

 View more versions of this product

Product Code. 15405227

  • £427.18 / 1000mL


Estimated Shipment date
from Supplier 07-08-2017
Add to basket

Specifications Description & Specifications

Specifications

1000mL
2° to 8°C
38°C (100°F)
Agarose Beads
9012-36-6
100°C
Documentation

Technical Documents