missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGAP3 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB30459.100uL
Additional Details : Weight : 0.02000kg
Description
Rabbit polyclonal antibody raised against partial recombinant human AGAP3.
- High specificity
- High reactivity
Specifications
AGAP3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human AGAP3. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
CENTG3/CRAG/FLJ16146/MRIP-1 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
116988 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20-1:50) The optimal working dilution should be determined by the end user. | |
Q96P47 | |
AGAP3 | |
Recombinant protein corresponding to human AGAP3. | |
100 μL | |
Primary | |
Human | |
Liquid |
For Research Use Only (RUO)