missing translation for 'onlineSavingsMsg'
Learn More

AG 825, Tocris Bioscience™

Selective ErbB2 inhibitor

Brand:  Tocris (Bio-Techne) 155550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13429673

  • £560.07 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective ErbB2 inhibitor (IC50 values are 0.15 and 19 μM at ErbB2 and ErbB1 respectively). Preferentially triggers p38 MAP kinase-dependent apoptosis in androgen-independent prostate cancer cells.
Specifications

Chemical Identifiers

149092-50-2
397.467
tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag
CHEBI:75405
COC1=CC(=CC(=C1O)CSC2=NC3=CC=CC=C3S2)C=C(C#N)C(=O)N
C19H15N3O3S2
KXDONFLNGBQLTN-WUXMJOGZSA-N
6091659
(E)-3-[3-(1,3-benzothiazol-2-ylsulfanylmethyl)-4-hydroxy-5-methoxyphenyl]-2-cyanoprop-2-enamide

Specifications

50mg
149092-50-2
tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag
COC1=CC(=CC(=C1O)CSC2=NC3=CC=CC=C3S2)C=C(C#N)C(=O)N
397.467
CHEBI:75405
>98%
AG 825
C19H15N3O3S2
KXDONFLNGBQLTN-WUXMJOGZSA-N
(E)-3-[3-(1,3-benzothiazol-2-ylsulfanylmethyl)-4-hydroxy-5-methoxyphenyl]-2-cyanoprop-2-enamide
6091659
397.47
Videos
Documents