missing translation for 'onlineSavingsMsg'
Learn More

AG 556, Tocris Bioscience™

EGFR-kinase inhibitor

Brand:  Tocris (Bio-Techne) 061610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13447813

  • £144.06 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Epidermal growth factor receptor (EGFR) kinase inhibitor (IC50 = 1.1 μM). Selective over ErbB2 (IC50 > 500 μM).
Specifications

Chemical Identifiers

133550-41-1
336.391
tyrphostin b56, e-2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl prop-2-enamide, 2e-2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl prop-2-enamide, tyrphostin ag 556, tyrphostin ag556, tyrphostin ag-556, tocris-0616, tyrphostin deriv. 56, 2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl acrylamide, alpha-cyano-3,4-dihydroxy-n-4-phenylbutyl cinnamide
(E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(4-phenylbutyl)prop-2-enamide
C20H20N2O3
GWCNJMUSWLTSCW-SFQUDFHCSA-N
5328775
C1=CC=C(C=C1)CCCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N

Specifications

10mg
133550-41-1
tyrphostin b56, e-2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl prop-2-enamide, 2e-2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl prop-2-enamide, tyrphostin ag 556, tyrphostin ag556, tyrphostin ag-556, tocris-0616, tyrphostin deriv. 56, 2-cyano-3-3,4-dihydroxyphenyl-n-4-phenylbutyl acrylamide, alpha-cyano-3,4-dihydroxy-n-4-phenylbutyl cinnamide
C1=CC=C(C=C1)CCCCNC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
336.391
336.39
AG 556
C20H20N2O3
GWCNJMUSWLTSCW-SFQUDFHCSA-N
(E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-(4-phenylbutyl)prop-2-enamide
5328775
>99%
Videos
Documents