Learn More
AG 494, Tocris Bioscience™
Potent EGFR-kinase inhibitor
Brand:  Tocris (Bio-Techne) 061910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487813
  • £128.25 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Potent inhibitor of epidermal growth factor receptor (EGFR) kinase (IC50 = 0.7 μM). Selective over ErbB2, PDGFR and insulin receptor kinase (IC50 values are 42, 6 and > 100 μM respectively).
Specifications
Chemical Identifiers
133550-35-3
280.283
(E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
C16H12N2O3
HKHOVJYOELRGMV-XYOKQWHBSA-N
5328771
Specifications
AG 494
133550-35-3
C16H12N2O3
HKHOVJYOELRGMV-XYOKQWHBSA-N
(E)-2-cyano-3-(3,4-dihydroxyphenyl)-N-phenylprop-2-enamide
5328771
10mg
>98%
(E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
C1=CC=C(C=C1)NC(=O)C(=CC2=CC(=C(C=C2)O)O)C#N
280.283
280.28
Videos