missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B2, crystalline, ACROS Organics™

Brand:  Acros Organics 227350010

UNSPSC : 12161500

Code : A0

Additional Details : CAS Number : 7220-81-7 Weight : 0.06000kg Transport : UN number : 3462 Chem class : 1 Pack group : I


Quantity 1mg

 View more versions of this product

Product Code. 10105150

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications Description & Specifications

Chemical Identifiers

Aflatoxin B2
C17H14O6
MFCD00078140
≥98%
314.28

Specifications

Aflatoxin B2
Authentic
≥98%
314.28
19, V,10, 552
Off-white
7220-81-7
Powder
C17H14O6
MFCD00078140
15, 171
1mg

Safety and Handling

Product Identifier
  • Aflatoxin B2

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 1A

Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

Documentation

Technical Documents