missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up

Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.

Brand:  Applied Biosystems™ 78205.10.ML

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15503687

  • £3400.00 / 5000 reactions
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
  • 30-minute protocol
Specifications

Specifications

-20°C
5000
10 mL
Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs
Single-step enzymatic PCR Cleanup Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers