missing translation for 'onlineSavingsMsg'
Learn More

AF-DX 116, Tocris Bioscience™

Selective M2 antagonist

Brand:  Tocris (Bio-Techne) 110550

Code : Z1

Additional Details : CAS Number : 102394-31-0 Weight : 0.00005kg

Quantity 50mg

 View more versions of this product

Product Code. 13428503

  • £635.04 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective M2 muscarinic receptor antagonist. Ki values are 64, 417, 786, 211 and 5130 nM for human recombinant M2, M1, M3, M4 and M5 muscarinic receptors, respectively.
Specifications

Chemical Identifiers

102394-31-0
421.545
otenzepad, af-dx 116, af-dx-116, unii-sx2wtg9r3z, unii-ww6a9tfl2c, sx2wtg9r3z, ww6a9tfl2c, chembl17045, 11-2-diethylamino methyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 11-2-2-diethylamino methyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
11-[2-[2-(diethylaminomethyl)piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
C24H31N5O2
UBRKDAVQCKZSPO-UHFFFAOYSA-N
107867
CCN(CC)CC1CCCCN1CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4

Specifications

50mg
102394-31-0
otenzepad, af-dx 116, af-dx-116, unii-sx2wtg9r3z, unii-ww6a9tfl2c, sx2wtg9r3z, ww6a9tfl2c, chembl17045, 11-2-diethylamino methyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 11-2-2-diethylamino methyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
CCN(CC)CC1CCCCN1CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4
421.545
421.54
AF-DX 116
C24H31N5O2
UBRKDAVQCKZSPO-UHFFFAOYSA-N
11-[2-[2-(diethylaminomethyl)piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
107867
>98%
Videos
Documents