missing translation for 'onlineSavingsMsg'
Learn More

AEG 3482, Tocris Bioscience™

Inhibitor of JNK signaling

Brand:  Tocris (Bio-Techne) 265110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13461654

  • £152.88 / 10mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of c-jun N-terminal kinase (JNK) signaling. Binds Hsp90 and facilitates HSF1 release, induces expression of Hsp70, which in turn blocks JNK activation and JNK-dependent apoptosis. Antiapoptotic; inhibits NGF withdrawal-induced death in SCG neurons (EC50 = 20 μM).
Specifications

Chemical Identifiers

63735-71-7
280.32
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
C10H8N4O2S2
MQUYTXDAVCOCMX-UHFFFAOYSA-N
698112
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N

Specifications

10mg
63735-71-7
unii-7ezf1a283n, 6-phenylimidazo 2,1-b 1,3,4 thiadiazole-2-sulfonamide, 6-phenylimidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, imidazo 2,1-b-1,3,4-thiadiazole-2-sulfonamide, 6-phenyl, chembl19231, aeg hplc, 6-phenylimidazo 2,1-b 1,3,4-thiadiazoline-2-sulfonamide
C1=CC=C(C=C1)C2=CN3C(=N2)SC(=N3)S(=O)(=O)N
280.32
280.33
AEG 3482
C10H8N4O2S2
MQUYTXDAVCOCMX-UHFFFAOYSA-N
6-phenylimidazo[2,1-b][1,3,4]thiadiazole-2-sulfonamide
698112
>99%
Videos
Documents