missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Packaging:
Poly Micro Tube
Description
This specific, irreversible inhibitor of serine proteases is a stable, nontoxic alternative to PMSF.
Specifications
Melting Point | 175°C |
Color | White |
Assay Percent Range | ≥98.0 % |
Packaging | Poly Micro Tube |
Quantity | 100 mg |
Formula Weight | 239.7 |
Percent Purity | ≥98 (HPLC) |
Physical Form | Powder or Solid |
Chemical Name or Material | AEBSF Hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'