missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Advantec™ All-Glass Microanalysis Filter Holder

Ideal for small to medium volumes of liquid

Brand:  Cole-Parmer™ 06645-13

Code : NEW

Additional Details : Weight : 0.24000kg

Product Code. 15331256

  • £279.51 / Each
Estimated Shipment: 20-05-2024
to see stock.
Add to basket

Description

Description

  • Microanalysis filter holder
  • Features borosilicate glass funnel and base, silicone rubber stopper and anodized aluminum clamp
  • 90mm holders are ideal for large-volume viscous samples or those with a heavy particle or bioburden that would clog a 47mm filter
Specifications

Specifications

Sintered Glass
All-Glass Microanalysis Filter Holder
15 mL
25mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers