missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protective Disposable Facemask, Certified High Quality
High quality CE marked Class I medical device facemasks, certified under EU Directive MDD 93/42/EEC and EN14683:2019 compliant. These latex-free polypropylene 3-ply pleated masks with earloops provide excellent fit with adjustable nose seal for extra security. Fully certified, quality guaranteed.
Brand: N/A 12888004
Description
- Disposable, single-use protection
- A CE marked; Class I medical device, certified under EU Directive MDD 93/42/EEC meeting the EN14683:2019 standard for medical facemasks
- Ear loops and a soft, flexible, internally wired upper seam for a comfortable snug fit around the nose
- Nonwoven polypropylene outer layer
- Thermally bonded nonwoven polypropylene inner layer
- Latex-free reducing the risks of skin irritation for the wearer
- Blue exterior
- Universal size and easy fitting
Specifications
Yes | |
CE marked; Class I medical device, certified under EU Directive MDD 93/42/EEC; Meets EN14683:2019 standard for Medical Face Masks | |
Non-woven Polypropylene (outer and inner), Meltblown Polypropylene (filter) | |
17.5 cm | |
Standard |
3-ply | |
Blue | |
Ear loops | |
Polypropylene | |
9.5 cm |