Learn More
ADSS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555070
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KLDGEIIPHI PANQEVLNKV EVQYKTLPGW NTDISNARAF KELPVNAQNY VRFIEDELQI PVKWIGVGKS RESM Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
ADSS Polyclonal antibody specifically detects ADSS in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ADSS | |
Polyclonal | |
Unconjugated | |
ADSS2 | |
ADEH; adenylosuccinate synthase; adenylosuccinate synthetase (Ade(-)H-complementing); adenylosuccinate synthetase 2, non muscle; adenylosuccinate synthetase isozyme 2; Adenylosuccinate synthetase, acidic isozyme; adenylosuccinate synthetase, liver isozyme; adenylosuccinate synthetase, non muscle; adenylosuccinate synthetase, non-muscle isozyme; Adss; ADSS 2; Adss2; AI314886; AMPSase 2; AS; IMP--aspartate ligase 2; L-type adenylosuccinate synthetase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P30520 | |
ADSS2 | |
Recombinant protein corresponding to Human ADSS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only