missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ ADSL Recombinant Human Protein, N-His tag

Expressed from DNA sequence encoding human ADSL (Met 1-Leu 484) fused to polyhistidine tag at N-terminus

Brand:  Sino Biological™ 11287H07E50

UNSPSC : 12352202

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208485

  • £311.00 / 50µg


Estimated Shipment
08-09-2017
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Formulation: PBS, 10% Glycerol, pH 7.4
Specifications

Specifications

Lyophilized
57kd
50μg
Recombinant
Adenylosuccinate Lyase
Assay Standard
E.coli
His Tag
158
≥90% by SDS-PAGE
Human
<1 EU/μg
ADSL
Other proteins
ADSL Recombinant Human Protein