missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADPGK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549873
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total ADPGK protein.
ADPGK Polyclonal antibody specifically detects ADPGK in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ADPGK | |
Polyclonal | |
Unconjugated | |
ADPGK | |
2610017G09Rik; ADP dependent glucokinase; ADP-dependent glucokinase; ADPGK; Adp-gk; ATP-dependent glucokinase; C77577; EC 2.7.1.147; PSEC0260; RbBP-35; RGD1306103 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
315722, 72141, 83440 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8VDL4, Q9BRR6 | |
ADPGK | |
Synthesized peptide derived from internal of human ADPGK. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |