missing translation for 'onlineSavingsMsg'
Learn More

Adiponitrile, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number p-7058964
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N
This item is not returnable. View return policy
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N

Adiponitrile is used as an intermediate for the synthesis of hexamethylenediamine (diamino-1,6-hexane). This chemical is an important intermediate for the manufacture of synthetic fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY
Melting Point 2°C
Density 0.951 g/mL
Boiling Point 295°C
Flash Point 163°C (325°F)
Odor Odorless
Packaging ChemSeal™ bottles
Refractive Index 1.438
Quantity 100 mL
UN Number UN2205
Beilstein 1740005
Solubility Information Highly soluble in water (83g/L).
Formula Weight 108.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Adiponitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'