Learn More
Thermo Scientific™ Adhesive PCR Plate Seals
Prevent sample evaporation during PCR, thermal cycling, or sample storage with this adhesive film.
£76.75 - £86.25
Specifications
For Use With PCR and qPCR
View More Specs
Products 3
Product Code Brand Description Temperature Range (Metric) No. per Pack Price Quantity & Availability  
Product Code Brand Description Temperature Range (Metric) No. per Pack Price Quantity & Availability  
11570274 View Documents Thermo Scientific™
AB0558
Peelable adhesive PCR film -20°F to +120°C 100 £82.50
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 24-01-2020
Add to basket
10130853 View Documents Thermo Scientific™
AB0626
Peelable adhesive PCR foil -40°C to +120°C 100 £76.75
Pack of 100 View alternative products
Save Up to 
»
EU Stock 378
Add to basket
12144741 View Documents Thermo Scientific™
AB1170
ABsolute qPCR Seal -80°C to +110°C 50 £86.25
Pack of 50 View alternative products
Save Up to 
»
EU Stock 88
Add to basket
Description
Description
Highlights
  • Clear for sample inspection
  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate when the seal is peeled off
  • End tabs for easy seal application and removal
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
Specifications
Specifications
PCR and qPCR