Learn More
ADH5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560761
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DYSFECIGNV KVMRAALEAC HKGWGVSVVV GVAASGEEIA TRPFQLVTGR TWKGTAFG Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
ADH5 Polyclonal antibody specifically detects ADH5 in Human samples. It is validated for Immunohistochemistry.Specifications
ADH5 | |
Polyclonal | |
Unconjugated | |
ADH5 | |
ADH1C; Adh2; Adh-2; ADH3; ADH-3; ADH5; Adh-5; ADH-B2; ADHX; alcohol dehydrogenase (class III), chi polypeptide; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2; Alcohol dehydrogenase 5; alcohol dehydrogenase 5 (class III), chi polypeptide; alcohol dehydrogenase B2; Alcohol dehydrogenase class chi chain; alcohol dehydrogenase class-3; Alcohol dehydrogenase class-III; class III alcohol dehydrogenase; class III alcohol dehydrogenase, chi subunit; epididymis secretory sperm binding protein Li 60p; FALDH; FDH; formaldehyde dehydrogenase; glutathione-dependent formaldehyde dehydrogenase; GSH-FDH; GSNOR; HEL-S-60p; S-(hydroxymethyl)glutathione dehydrogenase; S-nitrosoglutathione reductase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P11766 | |
ADH5 | |
Recombinant protein corresponding to Human ADH5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only