missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-monophosphate, 99%, Thermo Scientific Chemicals

Catalog Number p-3777172
27.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
61-19-8
C10H14N5O7P
347.22
MFCD00005750
UDMBCSSLTHHNCD-YPLCUDRINA-N
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
6083
CHEBI:16027
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
61-19-8
C10H14N5O7P
347.22
MFCD00005750
UDMBCSSLTHHNCD-YPLCUDRINA-N
adenosine 5'-monophosphate, 5'-adenylic acid, adenosine monophosphate, adenosine phosphate, adenylic acid, adenylate, phosphaden, 5'-amp, adenosine 5'-phosphate, phosphentaside
6083
CHEBI:16027
[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
NC1=C2N=CN([C@@H]3O[C@H](COP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.76 to 0.80 (A250/A260), 0.14 to 0.16 (A280/A260)
CAS Min % 98.5
CAS Max % 100.0
Melting Point 178.0°C to 185.0°C
Color Colorless to White
Loss on Drying 6% max. (105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 26, IV, 3615
Merck Index 15, 146
Solubility Information Solubility in water: soluble. Other solubilities: readily soluble in boiling water, soluble in ethanol
Spectrophotometry 98% min. (lambda max. 260nm,pH 7) molar absorptivity 15400+/-2%,on dry basis
Transmittance 95.0% min. (c=1, 0.2N NaOH, 430nm) 1cm cell
Formula Weight 347.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine 5′-monophosphate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'