missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine hydrochloride, 98+%, cont. up to ca 5% water

Catalog Number p-7023754
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
2922-28-3
C5H6ClN5
171.59
MFCD00038990
UQVDQSWZQXDUJB-UHFFFAOYSA-N
adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
76219
7H-purin-6-amine;hydrochloride
Cl.NC1=C2NC=NC2=NC=N1
This item is not returnable. View return policy
2922-28-3
C5H6ClN5
171.59
MFCD00038990
UQVDQSWZQXDUJB-UHFFFAOYSA-N
adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
76219
7H-purin-6-amine;hydrochloride
Cl.NC1=C2NC=NC2=NC=N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼290°C (decomposition)
Quantity 5 g
Beilstein 4009585
Formula Weight 171.59
Percent Purity ≥98%
Assay cont. up to ≈5% water
Chemical Name or Material Adenine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'