Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ TX-150 Swinging Bucket Rotors
Assorted tube adapters
Brand:  Thermo Scientific 75005736
Code : 03
Additional Details : Weight : 5.35690kg
 View more versions of this product
Product Code. 15184233
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
29.5mm
120mm
50mL Conical Buckets for TX-150 rotor
50mL Conical or Skirted Tube
Videos