missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters For Fixed-Angle Rotor
Add flexibility to Eppendorf Centrifuges with these rotor adapters
£80.55 - £133.00
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | Fixed angle rotor |
No. per Pack | 2 Pack |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10043061
|
Eppendorf™
5804770005 |
1.5 to 2 mL |
£133.00
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10428991
|
Eppendorf™
5804773004 |
20 to 30 mL |
£80.55
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Adapters
- For Eppendorf™ Fixed Angle Rotors
Specifications
Adapter | |
Fixed angle rotor | |
2 Pack |