missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters for Rectangular Buckets in A-4-81 Rotor
Add flexibility to A-4-81 Rotor for Eppendorf Centrifuge 5810/5810 R with these adapters
Brand: Eppendorf™ 5825722000
Code : A0
Additional Details : Weight : 1.00000kg
Description
- Rotor adapters
- For rectangluar buckets
- Available in a wide range of sizes to suit various applications
Specifications
Adapter | |
180 to 250 mL | |
62 mm | |
400mL rectangular bucket for use with standard, Vacutainer, Falcon tubes and flasks |
Standard, Vacutainer, Falcon Tubes and Flasks | |
500mL Rectangular Bucket for Rotor A-4-81 | |
1 | |
2 Pack |