Learn More
DWK Life Sciences DURAN™ Funnel Adapter
The filter crucible adapter with integrated stem serves to hold the filter funnel.
Brand:  DWK Life Sciences 243162206
Code : A0
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 11870700
  • £236.00 / Pack of 10
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Filter crucibles without stems can be extended with the filter crucible adapter. The integrated outlet stem allows a clean filling of the filtrate into the filtering flask.
  • Extension with integrated handle for receiving the filter crucible
  • Fired-on, highly durable white print
Specifications
Specifications
34 mm
Yes
10/Pk.
Boro 3.3
Separation of the filtrate for its further processing; Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering; Matching rubber adapter available