Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 6431
Code : 04
Additional Details : Weight : 0.13000kg
 View more versions of this product
Product Code. 10584922
  • £142.04 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M16 x 1 female to M30 x 1.5 male
Thermostat
Adapter