Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6278
Code : 04
Additional Details : Weight : 0.02430kg
Description
- Adapter
Specifications
Thread Size: M16 x 1 male to M16 x 1 male | |
Thermostat | |
Adapter | |
16 x 1 m |