Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand:  HUBER 5443
Code : 04
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10289551
  • £102.93 / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • Adapter
Specifications
Specifications
Thread Size: M16 x 1 male to G3/4 female
Thermostat
Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products