Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6454
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Adapter
Specifications
Thread Size: M16 x 1 male to M30 x 1.5 female | |
Thermostat | |
Adapter |