Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6394
Code : 04
Additional Details : Weight : 0.14000kg
Description
- Adapter
Specifications
Thread Size: M30 x 1.5 male to Rp1/2 female | |
Adapter |