missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ADAM23 Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | ADAM23 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
ADAM23 Polyclonal specifically detects ADAM23 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ADAM23 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
a disintegrin and metalloproteinase domain 23, ADAM metallopeptidase domain 23, disintegrin and metalloproteinase domain-containing protein 23, MDC3, MDC-3, MDC3ADAM 23, MDC-L, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3 | |
ADAM23 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
O75077 | |
8745 | |
Chinese hamster ovary cell line CHO-derived recombinant human ADAM23 Ser60-His585 Accession # O75077 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ADAM23 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) ADAM8, rhADAM9, and rhADAM22 is observed. |
For Research Use Only