missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ADAM19 Ectodomain Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | ADAM19 |
---|---|
Clone | 164646 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
ADAM19 Monoclonal specifically detects ADAM19 in Mouse samples. It is validated for Western Blot.Specifications
ADAM19 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
O35674 | |
8728 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM19 Ectodomain Asp205-Gly705 (predicted) Accession # O35674 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse ADAM19 Ectodomain in direct ELISAs and Western blots. It also recognizes a C-terminal fragment (amino acids 588-705) of recombinant mouse (rm) ADAM19 in Western blots. In Western blots, no cross-reactivity with rmADAM10 or rmADAM15 is observed. |
164646 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a disintegrin and metalloproteinase domain 19 (meltrin beta), ADAM 19, ADAM metallopeptidase domain 19, disintegrin and metalloproteinase domain-containing protein 19, EC 3.4.24, EC 3.4.24.-, MADDAM, Meltrin beta, Meltrin-beta, Metalloprotease and disintegrin dendritic antigen marker, MLTNBmeltrin beta | |
ADAM19 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only