missing translation for 'onlineSavingsMsg'
Learn More

ADA, 98+%, for biochemistry, ACROS Organics™

Brand:  Acros Organics 327681000

Code : 29

Additional Details : CAS Number : 26239-55-4 Weight : 0.15000kg

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10236051

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

ADA
98.0
For biochemistry
98+%
H2NCOCH2N(CH2CO2H)2
MFCD00008031
26239-55-4
100.0
≥98%
C6H10N2O5
190.16
N-(2-Acetamido)iminodiacetic acid

Specifications

ADA
98.0
Authentic
Plastic bottle
≥98%
C6H10N2O5
190.16
15, 135
Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform
White to almost white
100g
26239-55-4
100.0
For biochemistry
Powder
98+%
H2NCOCH2N(CH2CO2H)2
MFCD00008031
N-(2-Acetamido)iminodiacetic acid
1% max. (K.F.)
219.0°C
Videos
SDS